Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclopropyl-2-(1-piperazinyl)nicotinamide hydrochloride, 97%

$196.68 - $196.68

Chemical Identifiers

Chemical Name or Material N-Cyclopropyl-2-(1-piperazinyl)nicotinamide hydrochloride
CAS 902836-65-1
Molecular Formula C13H18N4O·HCl
Formula Weight 282.77g/mol
MDL Number MFCD12407085
Synonym 1-[3-(N-Cyclopropylcarbamoyl)-2-pyridyl]piperazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5168303

 
alfa aesar™
H51683-03
1g Each for $196.68
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material N-Cyclopropyl-2-(1-piperazinyl)nicotinamide hydrochloride
CAS 902836-65-1
Assay 97%
Molecular Formula C13H18N4O·HCl
Formula Weight 282.77g/mol
MDL Number MFCD12407085
Synonym 1-[3-(N-Cyclopropylcarbamoyl)-2-pyridyl]piperazine hydrochloride