Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclohexylmaleimide, 97%

$195.49 - $678.82

Chemical Identifiers

Chemical Name or Material N-Cyclohexylmaleimide
CAS 1631-25-0
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00043904
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2305606

 
alfa aesar™
B2305606
5g Each for $195.49

AAB2305614

 
alfa aesar™
B2305614
25g Each for $678.82
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material N-Cyclohexylmaleimide
TSCA TSCA
Assay 97%
Molecular Formula C10H13NO2
Melting Point 89° to 91°C
MDL Number MFCD00043904
CAS 1631-25-0
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.