Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclohexylethanolamine, 97%

$67.81 - $213.96

Chemical Identifiers

Chemical Name or Material N-Cyclohexylethanolamine
CAS 2842-38-8
Molecular Formula C8H17NO
Formula Weight 143.23
MDL Number MFCD00021326
Beilstein 1071413
Synonym 2-(Cyclohexylamino)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1082122

  SDS
alfa aesar™
L1082122
100g Each for $213.96

AAL1082114

  SDS
alfa aesar™
L1082114
25g Each for $67.81
Description & Specifications

Specifications

Chemical Name or Material N-Cyclohexylethanolamine
Boiling Point 112°C (3mmHg)
Melting Point 33° to 38°C
Quantity 100g
CAS 2842-38-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number KK3530000
Assay 97%
Molecular Formula C8H17NO
Formula Weight 143.23
MDL Number MFCD00021326
Beilstein 1071413
Synonym 2-(Cyclohexylamino)ethanol
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.