missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclohexylethanolamine, 97%

Manufacturer:  Alfa Aesar™ L1082122

 View more versions of this product

Catalog No. AAL1082122



Specifications

Specifications

N-Cyclohexylethanolamine
33°C to 38°C
2842-38-8
MFCD00021326
n-cyclohexylethanolamine, 2-cyclohexylamino ethanol, ethanol, 2-cyclohexylamino, abbomeen e-25 aerosol, cyclohexlaminoethanol, abromeen e-25, 2-cyclohexylamino ethan-1-ol, n-2-hydroxyethyl cyclohexylamine, cyclohexylamine, hydroxyethyl, n-2-hydroxyethyl cyclohexylamine
C1CCC(CC1)NCCO
143.23
143.23
112°C (3mmHg)
100g
C8H17NO
1071413
MGUMZJAQENFQKN-UHFFFAOYSA-N
2-(cyclohexylamino)ethanol
17827
97%

Chemical Identifiers

2842-38-8
143.23
MGUMZJAQENFQKN-UHFFFAOYSA-N
17827
C1CCC(CC1)NCCO
C8H17NO
MFCD00021326
n-cyclohexylethanolamine, 2-cyclohexylamino ethanol, ethanol, 2-cyclohexylamino, abbomeen e-25 aerosol, cyclohexlaminoethanol, abromeen e-25, 2-cyclohexylamino ethan-1-ol, n-2-hydroxyethyl cyclohexylamine, cyclohexylamine, hydroxyethyl, n-2-hydroxyethyl cyclohexylamine
2-(cyclohexylamino)ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : KK3530000

Documents