Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclohexylcarbodiimide, 0.8-1.0 mmol/g on Merrifield resin

Chemical Identifiers

Chemical Name or Material N-Cyclohexylcarbodiimide
Name Note 0.8 to 1.0mmol/g on Merrifield resin
MDL Number MFCD03410276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946303

  SDS
alfa aesar™
L1946303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946306

  SDS
alfa aesar™
L1946306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1946314

  SDS
alfa aesar™
L1946314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD03410276
Chemical Name or Material N-Cyclohexylcarbodiimide
Name Note 0.8 to 1.0mmol/g on Merrifield resin
Quantity 1g