missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclohexyl-N-methyl-4-nitrobenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 29477-27-8
Molecular Formula C14H18N2O3
Molecular Weight (g/mol) 262.309
MDL Number MFCD00709770
InChI Key ZGDQZPUGCAXWLR-UHFFFAOYSA-N
Synonym benzamide, n-cyclohexyl-n-methyl-4-nitro, aronis24032, n-cyclohexyl-n-methyl 4-nitrophenyl carboxamide
PubChem CID 692212
IUPAC Name N-cyclohexyl-N-methyl-4-nitrobenzamide
SMILES CN(C1CCCCC1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60410MD View Documents Alfa Aesar™
H60410MD
250mg Each for $220.53
 
AAH6041003 View Documents Alfa Aesar™
H6041003
1g Each for $703.20
 
Specifications

Chemical Identifiers

29477-27-8
262.309
ZGDQZPUGCAXWLR-UHFFFAOYSA-N
692212
CN(C1CCCCC1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]
C14H18N2O3
MFCD00709770
benzamide, n-cyclohexyl-n-methyl-4-nitro, aronis24032, n-cyclohexyl-n-methyl 4-nitrophenyl carboxamide
N-cyclohexyl-N-methyl-4-nitrobenzamide

Specifications

250mg
C14H18N2O3
benzamide, n-cyclohexyl-n-methyl-4-nitro, aronis24032, n-cyclohexyl-n-methyl 4-nitrophenyl carboxamide
CN(C1CCCCC1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]
262.309
262.3
N-Cyclohexyl-N-methyl-4-nitrobenzamide
29477-27-8
MFCD00709770
ZGDQZPUGCAXWLR-UHFFFAOYSA-N
N-cyclohexyl-N-methyl-4-nitrobenzamide
692212
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning