Please login to your online account to display your discounted pricing

N-Chlorosuccinimide, 98%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material N-Chlorosuccinimide
CAS 128-09-6
Molecular Formula C4H4ClNO2
Formula Weight 133.53
MDL Number MFCD00005511
UN Number UN3261
Beilstein 113915
Merck Index 14,2164
Synonym NCS
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA103100B

  SDS
alfa aesar
A103100B
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.53
Melting Point 146° to 150°C
Quantity 1000g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material N-Chlorosuccinimide
TSCA TSCA
Assay 98%
Molecular Formula C4H4ClNO2
MDL Number MFCD00005511
CAS 128-09-6
UN Number UN3261
Beilstein 113915
Merck Index 14,2164
Odor Chlorine-like
Synonym NCS
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number UY1013500

Applications
Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.

Solubility
Soluble in water, alcohol, benzene, acetone and acetic acid. Slightly soluble in ether, chloroform, carbon tetrachloride and petroleum ether.

Notes
Light sensitive and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, amines, ammonia, iron and iron salts.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.