Learn More
N-Chlorosuccinimide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1031030
Description
Regioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRegioselective chlorination and oxidizing reagentN-Chlorosuccinimide is used as a chlorinating agent, mild oxidant and swimming pool disinfectant. It is used in the microdetermination of hydrazine salts by titrimetry as well as in the detection of arsenic in the +3 oxidative state. It is also employed in the production of rubber additives. Further, it is used as pharmaceutical intermediate and antibiotic drugs intermediate.
Solubility
Soluble in water, alcohol, benzene, acetone and acetic acid. Slightly soluble in ether, chloroform, carbon tetrachloride and petroleum ether.
Notes
Light sensitive and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, amines, ammonia, iron and iron salts.
Specifications
N-Chlorosuccinimide, 98% | |
146°C to 150°C | |
98% | |
MFCD00005511 | |
113915 | |
n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 | |
JRNVZBWKYDBUCA-UHFFFAOYSA-N | |
1-chloropyrrolidine-2,5-dione | |
31398 | |
133.53 |
128-09-6 | |
1.65 g/mL | |
C4H4ClNO2 | |
250 g | |
Moisture sensitive | |
Soluble in water,alcohol,benzene,acetone and acetic acid. Slightly soluble in ether,chloroform,carbon tetrachloride and petroleum ether. | |
C1CC(=O)N(C1=O)Cl | |
133.531 | |
CHEBI:53203 |
Chemical Identifiers
128-09-6 | |
133.531 | |
JRNVZBWKYDBUCA-UHFFFAOYSA-N | |
31398 | |
1-chloropyrrolidine-2,5-dione |
C4H4ClNO2 | |
MFCD00005511 | |
n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 | |
CHEBI:53203 | |
C1CC(=O)N(C1=O)Cl |
Safety and Handling
P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
H302-H314-H335-H500
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 204-878-8
RTECSNumber : UY1013500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only