missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Chloromethyl)phthalimide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1201822

 View more versions of this product

Catalog No. AAA1201822



Description

Description

It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
Specifications

Specifications

N- (Chloromethyl)phthalimide
131°C to 135°C
MFCD00005898
140942
Insoluble in water.
C1=CC=C2C(=C1)C(=O)N(C2=O)CCl
195.602
195.61
17564-64-6
C9H6ClNO2
100 g
n-chloromethyl phthalimide, 2-chloromethyl isoindoline-1,3-dione, n-chloromethylphthalimide, chloromethylphthalimide, n-chloromethyltrimellitimide, 1h-isoindole-1,3 2h-dione, 2-chloromethyl, phthalimide, n-chloromethyl, 2-chloromethyl isoindole-1,3-dione, 2-chloromethyl-1h-isoindole-1,3 2h-dione
JKGLRGGCGUQNEX-UHFFFAOYSA-N
2-(chloromethyl)isoindole-1,3-dione
87154
97%

Chemical Identifiers

17564-64-6
195.602
JKGLRGGCGUQNEX-UHFFFAOYSA-N
87154
C1=CC=C2C(=C1)C(=O)N(C2=O)CCl
C9H6ClNO2
MFCD00005898
n-chloromethyl phthalimide, 2-chloromethyl isoindoline-1,3-dione, n-chloromethylphthalimide, chloromethylphthalimide, n-chloromethyltrimellitimide, 1h-isoindole-1,3 2h-dione, 2-chloromethyl, phthalimide, n-chloromethyl, 2-chloromethyl isoindole-1,3-dione, 2-chloromethyl-1h-isoindole-1,3 2h-dione
2-(chloromethyl)isoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 241-541-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only