missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Chloroacetylisonipecotic acid, 98%

$105.64 - $388.61

Chemical Identifiers

CAS 318280-69-2
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
MDL Number MFCD02093986
InChI Key PEQVHRULPWCLOJ-UHFFFAOYSA-N
Synonym n-chloroacetylisonipecotic acid, 1-2-chloroacetyl piperidine-4-carboxylic acid, 1-2-chloroacetyl-4-piperidinecarboxylic acid, acmc-20ak5i, n-chloroacetyl-dl-isonipecotic acid, 1-chloroacetylpiperidine-4-carboxylic acid, 1-chloroacetyl-4-piperidinecarboxylic acid, 1-chloroacetyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-chloroacetyl, 4-piperidinecarboxylic acid, 1-chloroacetyl-9ci
PubChem CID 1991140
IUPAC Name 1-(2-chloroacetyl)piperidine-4-carboxylic acid
SMILES C1CN(CCC1C(=O)O)C(=O)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1805206 View Documents Alfa Aesar™
L1805206
5g Each for $388.61
 
AAL1805203 View Documents Alfa Aesar™
L1805203
1g Each for $105.64
 
Specifications

Chemical Identifiers

318280-69-2
205.638
PEQVHRULPWCLOJ-UHFFFAOYSA-N
1991140
C1CN(CCC1C(=O)O)C(=O)CCl
C8H12ClNO3
MFCD02093986
n-chloroacetylisonipecotic acid, 1-2-chloroacetyl piperidine-4-carboxylic acid, 1-2-chloroacetyl-4-piperidinecarboxylic acid, acmc-20ak5i, n-chloroacetyl-dl-isonipecotic acid, 1-chloroacetylpiperidine-4-carboxylic acid, 1-chloroacetyl-4-piperidinecarboxylic acid, 1-chloroacetyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-chloroacetyl, 4-piperidinecarboxylic acid, 1-chloroacetyl-9ci
1-(2-chloroacetyl)piperidine-4-carboxylic acid

Specifications

109°C to 111°C
318280-69-2
MFCD02093986
PEQVHRULPWCLOJ-UHFFFAOYSA-N
1-(2-chloroacetyl)piperidine-4-carboxylic acid
1991140
98%
5g
C8H12ClNO3
n-chloroacetylisonipecotic acid, 1-2-chloroacetyl piperidine-4-carboxylic acid, 1-2-chloroacetyl-4-piperidinecarboxylic acid, acmc-20ak5i, n-chloroacetyl-dl-isonipecotic acid, 1-chloroacetylpiperidine-4-carboxylic acid, 1-chloroacetyl-4-piperidinecarboxylic acid, 1-chloroacetyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-chloroacetyl, 4-piperidinecarboxylic acid, 1-chloroacetyl-9ci
C1CN(CCC1C(=O)O)C(=O)CCl
205.638
205.64
N-Chloroacetylisonipecotic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning