missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Chloroacetyl-L-tyrosine 98.0+%, TCI America™

Manufacturer:  TCI America C01311G

 View more versions of this product

Catalog No. C01311G



Specifications

Specifications

N-Chloroacetyl-L-tyrosine
153°C
1145-56-8
MFCD00002390
GDOGSOZOUAVIFX-VIFPVBQESA-N
(2S)-2-[(2-chloroacetyl)amino]-3-(4-hydroxyphenyl)propanoic acid
2724332
Crystal-Powder at 20°C
White-Yellow
1g
C11H12ClNO4
n-chloroacetyl-l-tyrosine, chloroacetyl-l-tyrosine, s-2-2-chloroacetamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2-chloroacetamido-3-4-hydroxyphenyl propanoic acid, chloracetyltyrosin, chloroacetyl-l-tyr-oh, l-tyrosine, n-chloroacetyl, n-2-chloroacetyl-l-tyrosine, n-chloroacetyl-4-hydroxyphenylalanine #, 2s-2-2-chloroacetylamino-3-4-hydroxyphenyl propanoic acid
C1=CC(=CC=C1CC(C(=O)O)NC(=O)CCl)O
257.67
257.67
≥98.0% (T)

Chemical Identifiers

1145-56-8
257.67
GDOGSOZOUAVIFX-VIFPVBQESA-N
2724332
C1=CC(=CC=C1CC(C(=O)O)NC(=O)CCl)O
C11H12ClNO4
MFCD00002390
n-chloroacetyl-l-tyrosine, chloroacetyl-l-tyrosine, s-2-2-chloroacetamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2-chloroacetamido-3-4-hydroxyphenyl propanoic acid, chloracetyltyrosin, chloroacetyl-l-tyr-oh, l-tyrosine, n-chloroacetyl, n-2-chloroacetyl-l-tyrosine, n-chloroacetyl-4-hydroxyphenylalanine #, 2s-2-2-chloroacetylamino-3-4-hydroxyphenyl propanoic acid
(2S)-2-[(2-chloroacetyl)amino]-3-(4-hydroxyphenyl)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents