Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Chloroacetyl)glycine, 98+%

$69.27 - $267.84

Chemical Identifiers

Chemical Name or Material N-(Chloroacetyl)glycine
CAS 6319-96-6
Molecular Formula ClCH2CONHCH2CO2H
Formula Weight 151.55
MDL Number MFCD00021733
Beilstein 1765217
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0765106

  SDS
alfa aesar™
L0765106
5g Each for $69.27

AAL0765114

  SDS
alfa aesar™
L0765114
25g Each for $267.84
Description & Specifications

Specifications

Formula Weight 151.55
Chemical Name or Material N-(Chloroacetyl)glycine
Assay ≥98%
CAS 6319-96-6
Molecular Formula ClCH2CONHCH2CO2H
MDL Number MFCD00021733
Beilstein 1765217
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 98° to 106°C
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.