missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-CBZ-D-glutamic acid α-benzyl ester, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440940050
Specifications
N-CBZ-D-glutamic acid α-benzyl ester | |
24 | |
White | |
5g | |
97.5 | |
97.5% min. (HPLC) | |
1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, cbz-d-glu oh-obzl, cbz-d-glu-obzl, cbz-d-glu-och2ph, n-cbz-d-glutamic acid alpha-benzyl ester, pubchem14966, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, z-d-glu-obzl | |
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2 | |
371.39 | |
371.39 | |
98% |
Authentic | |
1% Max. | |
97°C to 98°C | |
65706-99-2 | |
100.0 | |
C20H21NO6 | |
VWHKODOUMSMUAF-QGZVFWFLSA-N | |
(4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid | |
7018162 | |
Powder |
Chemical Identifiers
65706-99-2 | |
371.39 | |
1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, cbz-d-glu oh-obzl, cbz-d-glu-obzl, cbz-d-glu-och2ph, n-cbz-d-glutamic acid alpha-benzyl ester, pubchem14966, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, z-d-glu-obzl | |
(4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid |
C20H21NO6 | |
VWHKODOUMSMUAF-QGZVFWFLSA-N | |
7018162 | |
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2 |