missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-CBZ-D-glutamic acid α-benzyl ester, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440940010

 View more versions of this product

Catalog No. AC440940010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-CBZ-D-glutamic acid α-benzyl ester
Glass Bottle
1% Max.
97°C to 98°C
65706-99-2
100.0
C20H21NO6
VWHKODOUMSMUAF-QGZVFWFLSA-N
(4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
7018162
Powder
Authentic
24
White
1g
97.5
97.5% min. (HPLC)
1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, cbz-d-glu oh-obzl, cbz-d-glu-obzl, cbz-d-glu-och2ph, n-cbz-d-glutamic acid alpha-benzyl ester, pubchem14966, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, z-d-glu-obzl
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2
371.39
371.39
98%

Chemical Identifiers

65706-99-2
371.39
1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, cbz-d-glu oh-obzl, cbz-d-glu-obzl, cbz-d-glu-och2ph, n-cbz-d-glutamic acid alpha-benzyl ester, pubchem14966, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, z-d-glu-obzl
(4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
C20H21NO6
VWHKODOUMSMUAF-QGZVFWFLSA-N
7018162
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2
SDS Documents