missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Carbobenzyloxy-L-proline, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 1148-11-4 |
---|---|
Molecular Formula | C13H15NO4 |
Molecular Weight (g/mol) | 249.27 |
MDL Number | MFCD00003170 |
InChI Key | JXGVXCZADZNAMJ-NSHDSACASA-N |
Synonym | z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline |
PubChem CID | 101987 |
IUPAC Name | (2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
SMILES | C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC108370050
|
Thermo Scientific Chemicals
108370050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC108370100
|
Thermo Scientific Chemicals
108370100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1148-11-4 | |
249.27 | |
JXGVXCZADZNAMJ-NSHDSACASA-N | |
101987 | |
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
C13H15NO4 | |
MFCD00003170 | |
z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline | |
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
Specifications
1148-11-4 | |
100.0 | |
0.5% max. D-enantiomer (HPLC) | |
C13H15NO4 | |
5g | |
-60 | |
Solubility in water: negligible. Other solubilities: soluble in most common organic solvents | |
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
249.27 | |
249.27 | |
N-Carbobenzyloxy-L-proline, 99% |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00003170 | |
− 60.00 (20.00°C c=2,AcOH) | |
z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline | |
JXGVXCZADZNAMJ-NSHDSACASA-N | |
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid | |
101987 | |
99% |
Safety and Handling
EINECSNumber : 214-557-4
RTECSNumber : UY0745000
RUO â Research Use Only