missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Carbobenzyloxy-L-proline, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 108370100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Carbobenzyloxy-L-proline | |
1148-11-4 | |
100.0 | |
0.5% max. D-enantiomer (HPLC) | |
C13H15NO4 | |
10g | |
z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline | |
JXGVXCZADZNAMJ-NSHDSACASA-N | |
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid | |
101987 | |
99% |
99% | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00003170 | |
-60 | |
Solubility in water: negligible. Other solubilities: soluble in most common organic solvents | |
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O | |
249.27 | |
249.27 |
Chemical Identifiers
1148-11-4 | |
249.27 | |
JXGVXCZADZNAMJ-NSHDSACASA-N | |
101987 | |
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O |
C13H15NO4 | |
MFCD00003170 | |
z-pro-oh, n-benzyloxycarbonyl-l-proline, n-carbobenzoxy-l-proline, n-cbz-l-proline, benzyloxycarbonyl-l-proline, carbobenzoxyproline, carbobenzyloxy-l-proline, carbobenzoxy-l-proline, benzyloxycarbonylproline, n-carbobenzyloxy-l-proline | |
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid |
Safety and Handling
EINECSNumber : 214-557-4
RTECSNumber : UY0745000
RUO â Research Use Only