missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-L-methionine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442610250

 View more versions of this product

Catalog No. AC442610250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-Carbobenzyloxy-L-methionine
-27
67°C to 70°C
1152-62-1
100.0
C13H17NO4S
FPKHNNQXKZMOJJ-NSHDSACASA-N
(2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
1550857
Powder or Crystalline Powder
Authentic
White
25g
96.0
96% min. (HPLC)
benzyloxycarbonyl-l-methionine, cbz-l-methionine, cbz-met-oh, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine, n-carbobenzoxy-l-methionine, n-carbobenzyloxy-l-methionine, n-cbz-l-methionine, z-l-methionine, z-met-oh
CSCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
283.35
283.35
97%

Chemical Identifiers

1152-62-1
283.35
benzyloxycarbonyl-l-methionine, cbz-l-methionine, cbz-met-oh, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine, n-carbobenzoxy-l-methionine, n-carbobenzyloxy-l-methionine, n-cbz-l-methionine, z-l-methionine, z-met-oh
(2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
C13H17NO4S
FPKHNNQXKZMOJJ-NSHDSACASA-N
1550857
CSCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
Safety and Handling

Safety and Handling

EINECSNumber : 214-570-5

SDS Documents