missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-L-methionine, 97%, ACROS Organics™

Chemical Identifiers

CAS 1152-62-1
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.34
MDL Number MFCD00065124
InChI Key FPKHNNQXKZMOJJ-UHFFFAOYNA-N
Synonym benzyloxycarbonyl-l-methionine, cbz-l-methionine, cbz-met-oh, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine, n-carbobenzoxy-l-methionine, n-carbobenzyloxy-l-methionine, n-cbz-l-methionine, z-l-methionine, z-met-oh
PubChem CID 1550857
IUPAC Name (2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442610050
View Documents
Thermo Fisher Scientific
AC442610050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442610250
SDS View Documents
Thermo Fisher Scientific
AC442610250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1152-62-1
283.34
FPKHNNQXKZMOJJ-UHFFFAOYNA-N
1550857
CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
C13H17NO4S
MFCD00065124
benzyloxycarbonyl-l-methionine, cbz-l-methionine, cbz-met-oh, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine, n-carbobenzoxy-l-methionine, n-carbobenzyloxy-l-methionine, n-cbz-l-methionine, z-l-methionine, z-met-oh
(2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

Authentic
White
5g
96.0
96% min. (HPLC)
MFCD00065124
FPKHNNQXKZMOJJ-UHFFFAOYNA-N
(2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
1550857
Powder or Crystalline Powder
N-Carbobenzyloxy-L-methionine
-27
67°C to 70°C
1152-62-1
100.0
C13H17NO4S
benzyloxycarbonyl-l-methionine, cbz-l-methionine, cbz-met-oh, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine, n-carbobenzoxy-l-methionine, n-carbobenzyloxy-l-methionine, n-cbz-l-methionine, z-l-methionine, z-met-oh
CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
283.34
283.35
97%
Safety and Handling

Safety and Handling

EINECSNumber : 214-570-5

SDS
Product Certifications