missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-ε-Carbobenzyloxy-L-lysine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC108360250

 View more versions of this product

Catalog No. AC108360250


 

Specifications Description & Specifications

Specifications

Nε-Carbobenzyloxy-L-lysine
97.5
Powder
98%
C14H20N2O4
280.32
Glass bottle
+14.0° to +16.0° (20°C, 589nm) (c=2, 1M HCl)
0.5% max. (K.F.)
White to off-white
1155-64-2
100.0
≥97.5%
Authentic
C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
MFCD00002638
Nε-CBZ-L-Lysine
0.2% max.
+15.50 (20.00°C c=1,1N HCl)
25g

Chemical Identifiers

1155-64-2
100.0
C14H20N2O4
280.32
Nε-CBZ-L-Lysine
≥97.5%
97.5
98%
C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
MFCD00002638
Nε-Carbobenzyloxy-L-lysine