Please login to your online account to display your discounted pricing

N-ε-Carbobenzyloxy-L-lysine, 98%, ACROS Organics™

$174.93 - $174.93

Chemical Identifiers

Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
CAS 1155-64-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H20N2O4
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
Formula Weight 280.32
MDL Number MFCD00002638
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC108360250

  SDS
thermo fisher scientific
AC108360250
Glass bottle 25g Each for $174.93

AC108360050

  SDS
fisher scientific acros
108360050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC108360010

  SDS
thermo fisher scientific
AC108360010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 280.32
Quantity 25g
Color White to off-white
Specific Rotation Condition +15.50 (20.00°C c=1,1N HCl)
Assay Percent Range 98%
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
CAS 1155-64-2
Synonym Nε-CBZ-L-Lysine
Sulfated Ash 0.2% max.
Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
Molecular Formula C14H20N2O4
MDL Number MFCD00002638
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +14.0° to +16.0° (20°C, 589nm) (c=2, 1M HCl)
Water 0.5% max. (K.F.)
CAS Max % 100.0