Please login to your online account to display your discounted pricing

N-ε-Carbobenzyloxy-L-lysine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  108360050

 View more versions of this product

Catalog No. AC108360050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 280.32
Physical Form Powder
Quantity 5g
Color White to off-white
Synonym Nε-CBZ-L-Lysine
Specific Rotation +14.0° to +16.0° (20°C, 589nm) (c=2, 1M HCl)
Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
Molecular Formula C14H20N2O4
MDL Number MFCD00002638
CAS 1155-64-2
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
Sulfated Ash 0.2% max.
Water 0.5% max. (K.F.)

Specifications

Chemical Name or Material Nε-Carbobenzyloxy-L-lysine
CAS 1155-64-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H20N2O4
Linear Formula C6H5CH2O2CNH(CH2)4CH(NH2)CO2H
Formula Weight 280.32
MDL Number MFCD00002638
Synonym Nε-CBZ-L-Lysine