Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-L-leucine, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  275970050

 View more versions of this product

Catalog No. AC275970050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 265.3
Physical Form Oil or Weak Solid
Quantity 5g
Color Yellow
EINECS Number 217-960-3
RTECS Number OH2921000
CAS Min % 94.0
CAS Max % 100.0
CAS 2018-66-8
Synonym N-CBZ-L-Leucine
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula CH(CH3)2CH2CH(COOH)NHC(O)OCH2C6H5
Chemical Name or Material N-Carbobenzyloxy-L-leucine
Molecular Formula C14H19NO4
Name Note 95%
MDL Number MFCD00026494
Packaging Glass bottle
Specific Rotation -16° to -18° (20°C, 589nm) (c=2, EtOH)

Specifications

Chemical Name or Material N-Carbobenzyloxy-L-leucine
Name Note 95%
CAS 2018-66-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H19NO4
Linear Formula CH(CH3)2CH2CH(COOH)NHC(O)OCH2C6H5
Formula Weight 265.3
MDL Number MFCD00026494
Synonym N-CBZ-L-Leucine