Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-L-leucine, 95%, ACROS Organics™

$33.82 - $33.82

Chemical Identifiers

Chemical Name or Material N-Carbobenzyloxy-L-leucine
Name Note 95%
CAS 2018-66-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H19NO4
Linear Formula CH(CH3)2CH2CH(COOH)NHC(O)OCH2C6H5
Formula Weight 265.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC275970050

  SDS
thermo fisher scientific
275970050
Glass bottle 5g Each for $33.82
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 265.3
Physical Form Oil or Weak Solid
Quantity 5g
Color Yellow
EINECS Number 217-960-3
RTECS Number OH2921000
CAS Min % 94.0
CAS Max % 100.0
CAS 2018-66-8
Synonym N-CBZ-L-Leucine
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula CH(CH3)2CH2CH(COOH)NHC(O)OCH2C6H5
Chemical Name or Material N-Carbobenzyloxy-L-leucine
Molecular Formula C14H19NO4
Name Note 95%
MDL Number MFCD00026494
Packaging Glass bottle
Specific Rotation -16° to -18° (20°C, 589nm) (c=2, EtOH)