missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-D-proline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC229520050

 View more versions of this product

Catalog No. AC229520050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Carbobenzyloxy-D-proline
+ 40.20 (22.00°C c=2,alcohol)
White
5g
97.5
97.5% min. (HPLC)
+-z-d-proline, 1-benzyloxy carbonyl-d-proline, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, n-benzyloxycarbonyl-d-proline, n-carbobenzoxy-d-proline, n-cbz-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, r-n-benzyloxycarbonylproline, z-d-pro-oh, z-d-proline
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
249.27
249.27
98%
Authentic
+ 40.20
74.6°C to 74.3°C
6404-31-5
100.0
C13H15NO4
JXGVXCZADZNAMJ-LLVKDONJSA-N
(2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
111208
Crystalline Powder or Powder

Chemical Identifiers

6404-31-5
249.27
+-z-d-proline, 1-benzyloxy carbonyl-d-proline, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, n-benzyloxycarbonyl-d-proline, n-carbobenzoxy-d-proline, n-cbz-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, r-n-benzyloxycarbonylproline, z-d-pro-oh, z-d-proline
(2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C13H15NO4
JXGVXCZADZNAMJ-LLVKDONJSA-N
111208
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : 229-021-5

SDS Documents