Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-D-proline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Carbobenzyloxy-D-proline
CAS 6404-31-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H15NO4
Formula Weight 249.27
Synonym N-CBZ-D-Proline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC229520050

 
thermo fisher scientific
AC229520050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC229520010

 
thermo fisher scientific
AC229520010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.27
Physical Form Crystalline Powder or Powder
Assay ≥97.5%
Quantity 5g
Color White
Specific Rotation Condition + 40.20 (22.00°C c=2,alcohol)
Melting Point 73° to 78°C
CAS Max % 100.0
Chemical Name or Material N-Carbobenzyloxy-D-proline
Specific Rotation +38° to +42° (20°C, 589nm) (c=2 in ethanol)
Assay Percent Range 98%
CAS Min % 97.5
CAS 6404-31-5
Molecular Formula C13H15NO4
Synonym N-CBZ-D-Proline
Infrared Spectrum Authentic