missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Carbobenzyloxy-D-alanine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441970050
Specifications
N-Carbobenzyloxy-D-alanine | |
1.00% max. () | |
White | |
5g | |
97.5 | |
97.5% min. (HPLC) | |
benzyloxycarbonyl-d-alanine, cbz-d-ala-oh, cbz-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, n-carbobenzyloxy-d-alanine, n-cbz-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, z-d-ala-oh, z-d-alanine | |
CC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
223.23 | |
223.23 | |
98% |
Authentic | |
+ 15.3 | |
74°C to 80°C | |
26607-51-2 | |
100.0 | |
C11H13NO4 | |
TYRGLVWXHJRKMT-MRVPVSSYSA-N | |
(2R)-2-(phenylmethoxycarbonylamino)propanoic acid | |
70832 | |
Crystalline Powder |
Chemical Identifiers
26607-51-2 | |
223.23 | |
benzyloxycarbonyl-d-alanine, cbz-d-ala-oh, cbz-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, n-carbobenzyloxy-d-alanine, n-cbz-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, z-d-ala-oh, z-d-alanine | |
(2R)-2-(phenylmethoxycarbonylamino)propanoic acid |
C11H13NO4 | |
TYRGLVWXHJRKMT-MRVPVSSYSA-N | |
70832 | |
CC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |