missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-D-alanine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441970010

 View more versions of this product

Catalog No. AC441970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Carbobenzyloxy-D-alanine
1.00% max. ()
+ 15.3
74°C to 80°C
26607-51-2
100.0
C11H13NO4
TYRGLVWXHJRKMT-MRVPVSSYSA-N
(2R)-2-(phenylmethoxycarbonylamino)propanoic acid
70832
Crystalline Powder
Authentic
Glass Bottle
White
1g
97.5
97.5% min. (HPLC)
benzyloxycarbonyl-d-alanine, cbz-d-ala-oh, cbz-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, n-carbobenzyloxy-d-alanine, n-cbz-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, z-d-ala-oh, z-d-alanine
CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
223.23
223.23
98%

Chemical Identifiers

26607-51-2
223.23
benzyloxycarbonyl-d-alanine, cbz-d-ala-oh, cbz-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, n-carbobenzyloxy-d-alanine, n-cbz-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, z-d-ala-oh, z-d-alanine
(2R)-2-(phenylmethoxycarbonylamino)propanoic acid
C11H13NO4
TYRGLVWXHJRKMT-MRVPVSSYSA-N
70832
CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
SDS Documents