Please login to your online account to display your discounted pricing

N-Carbobenzyloxy-D-alanine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Carbobenzyloxy-D-alanine
CAS 26607-51-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H13NO4
Formula Weight 223.23
Synonym N-CBZ-D-alanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441970010

 
thermo fisher scientific
AC441970010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441970050

 
thermo fisher scientific
AC441970050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 223.23
Quantity 1g
Color White to off-white
CAS Max % 100.0
Specific Rotation +14.0° to +16.0° (20°C, 589nm) (c=2, AcOH)
CAS 26607-51-2
Molecular Formula C11H13NO4
Assay Percent Range 98%
Chemical Name or Material N-Carbobenzyloxy-D-alanine
CAS Min % 97.5
Synonym N-CBZ-D-alanine
Loss on Drying 1.00% max. ()
Infrared Spectrum Authentic