missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-L-prolinamide, TCI America™

Manufacturer:  TCI America C300525G

 View more versions of this product

Catalog No. C300525G



Specifications

Specifications

N-Carbobenzoxy-L-prolinamide
92°C
34079-31-7
MFCD00020830
ZCGHEBMEQXMRQL-NSHDSACASA-N
benzyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
6950961
Crystal-Powder at 20°C
White-Yellow
25g
C13H16N2O3
z-pro-nh2, cbz-l-prolinamide, s-1-n-cbz-prolinamide, s-benzyl 2-carbamoylpyrrolidine-1-carboxylate, cbz-pro-nh2, z-l-prolinamide, z-l-proline amide, carbobenzyloxy-l-prolinamide, benzyl 2s-2-carbamoylpyrrolidine-1-carboxylate, benzyloxycarbonyl-l-proline amide
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)N
248.282
248.28
≥98.0% (HPLC,N)

Chemical Identifiers

34079-31-7
248.282
ZCGHEBMEQXMRQL-NSHDSACASA-N
6950961
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)N
C13H16N2O3
MFCD00020830
z-pro-nh2, cbz-l-prolinamide, s-1-n-cbz-prolinamide, s-benzyl 2-carbamoylpyrrolidine-1-carboxylate, cbz-pro-nh2, z-l-prolinamide, z-l-proline amide, carbobenzyloxy-l-prolinamide, benzyl 2s-2-carbamoylpyrrolidine-1-carboxylate, benzyloxycarbonyl-l-proline amide
benzyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents