missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-L-aspartic Acid, TCI America™

$22.92 - $62.64

Chemical Identifiers

CAS 1152-61-0
Molecular Formula C12H13NO6
Molecular Weight (g/mol) 267.237
MDL Number MFCD00002719
InChI Key XYXYXSKSTZAEJW-VIFPVBQESA-N
Synonym z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
PubChem CID 2723942
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C06295G View Documents TCI America
C06295G
5g Each for $22.92
 
C062925G View Documents TCI America
C062925G
25g Each for $62.64
 
Specifications

Chemical Identifiers

1152-61-0
267.237
XYXYXSKSTZAEJW-VIFPVBQESA-N
2723942
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
C12H13NO6
MFCD00002719
z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
(2S)-2-(phenylmethoxycarbonylamino)butanedioic acid

Specifications

White
5g
C12H13NO6
z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
267.237
267.24
≥99.0% (T)
116°C
1152-61-0
MFCD00002719
XYXYXSKSTZAEJW-VIFPVBQESA-N
(2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
2723942
Crystal-Powder at 20°C
N-Carbobenzoxy-L-aspartic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1549

TSCA : Yes