missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-DL-tryptophan, TCI America™

$47.36 - $132.92

Chemical Identifiers

CAS 13058-16-7
Molecular Formula C19H18N2O4
Molecular Weight (g/mol) 338.363
MDL Number MFCD00069707
InChI Key AHYFYYVVAXRMKB-UHFFFAOYSA-N
Synonym N-Cbz-DL-tryptophan, Z-DL-Trp-OH
PubChem CID 98198
IUPAC Name 3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CNC3=CC=CC=C32)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C06411G View Documents TCI America
C06411G
1g Each for $47.36
 
C06415G View Documents TCI America
C06415G
5g Each for $132.92
 
Specifications

Chemical Identifiers

13058-16-7
338.363
AHYFYYVVAXRMKB-UHFFFAOYSA-N
98198
C1=CC=C(C=C1)COC(=O)NC(CC2=CNC3=CC=CC=C32)C(=O)O
C19H18N2O4
MFCD00069707
N-Cbz-DL-tryptophan, Z-DL-Trp-OH
3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

Undesignated
1g
C19H18N2O4
N-Cbz-DL-tryptophan, Z-DL-Trp-OH
C1=CC=C(C=C1)COC(=O)NC(CC2=CNC3=CC=CC=C32)C(=O)O
338.363
338.36
≥97.0% (T)
170°C
13058-16-7
MFCD00069707
AHYFYYVVAXRMKB-UHFFFAOYSA-N
3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
98198
Crystal-Powder at 20°C
N-Carbobenzoxy-DL-tryptophan
Safety and Handling

Safety and Handling

TSCA : No