missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-DL-norvaline, TCI America™

$87.08 - $261.25

Chemical Identifiers

CAS 21691-43-0
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.282
MDL Number MFCD00020434
InChI Key NSJDRLWFFAWSFP-UHFFFAOYSA-N
Synonym z-dl-nva-oh, n-carbobenzoxy-dl-norvaline, n-cbz-dl-norvaline, n-benzyloxy carbonyl norvaline, n-cbz-l-norvaline, 2-benzyloxy carbonyl amino pentanoic acid, 2-phenylmethoxycarbonylamino pentanoic acid, dl-norvaline, n-phenylmethoxy carbonyl, z-dl-norvaline, cbz-dl-norvaline
PubChem CID 263473
IUPAC Name 2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C07525G View Documents TCI America
C07525G
5g Each for $87.08
 
C075225G View Documents TCI America
C075225G
25g Each for $261.25
 
Specifications

Chemical Identifiers

21691-43-0
251.282
NSJDRLWFFAWSFP-UHFFFAOYSA-N
263473
CCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C13H17NO4
MFCD00020434
z-dl-nva-oh, n-carbobenzoxy-dl-norvaline, n-cbz-dl-norvaline, n-benzyloxy carbonyl norvaline, n-cbz-l-norvaline, 2-benzyloxy carbonyl amino pentanoic acid, 2-phenylmethoxycarbonylamino pentanoic acid, dl-norvaline, n-phenylmethoxy carbonyl, z-dl-norvaline, cbz-dl-norvaline
2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

White
5g
C13H17NO4
z-dl-nva-oh, n-carbobenzoxy-dl-norvaline, n-cbz-dl-norvaline, n-benzyloxy carbonyl norvaline, n-cbz-l-norvaline, 2-benzyloxy carbonyl amino pentanoic acid, 2-phenylmethoxycarbonylamino pentanoic acid, dl-norvaline, n-phenylmethoxy carbonyl, z-dl-norvaline, cbz-dl-norvaline
CCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
251.282
251.28
≥98.0% (T)
88°C
21691-43-0
MFCD00020434
NSJDRLWFFAWSFP-UHFFFAOYSA-N
2-(phenylmethoxycarbonylamino)pentanoic acid
263473
Crystal-Powder at 20°C
N-Carbobenzoxy-DL-norvaline
Safety and Handling

Safety and Handling

TSCA : No