missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-DL-leucine, TCI America™

$45.83 - $333.06

Chemical Identifiers

CAS 3588-60-1
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00065129
InChI Key USPFMEKVPDBMCG-UHFFFAOYSA-N
Synonym N-Cbz-DL-leucine, Z-DL-Leu-OH
PubChem CID 246673
IUPAC Name 4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C06421G View Documents TCI America
C06421G
1g Each for $45.83
 
C06425G View Documents TCI America
C06425G
5g Each for $113.06
 
C064225G View Documents TCI America
C064225G
25g Each for $333.06
 
Specifications

Chemical Identifiers

3588-60-1
265.309
USPFMEKVPDBMCG-UHFFFAOYSA-N
246673
CC(C)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C14H19NO4
MFCD00065129
N-Cbz-DL-leucine, Z-DL-Leu-OH
4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

White
1g
C14H19NO4
N-Cbz-DL-leucine, Z-DL-Leu-OH
CC(C)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
265.309
265.31
≥98.0% (HPLC)
54°C
3588-60-1
MFCD00065129
USPFMEKVPDBMCG-UHFFFAOYSA-N
4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
246673
Crystal-Powder at 20°C
N-Carbobenzoxy-DL-leucine
Safety and Handling

Safety and Handling

TSCA : No