missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Carbobenzoxy-D-alanine 98.0+%, TCI America™
$26.54 - $189.13
Chemical Identifiers
CAS | 26607-51-2 |
---|---|
Molecular Formula | C11H13NO4 |
Molecular Weight (g/mol) | 223.23 |
MDL Number | MFCD00063126 |
InChI Key | TYRGLVWXHJRKMT-UHFFFAOYNA-N |
Synonym | n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine |
PubChem CID | 70832 |
IUPAC Name | 2-{[(benzyloxy)carbonyl]amino}propanoic acid |
SMILES | CC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
26607-51-2 | |
223.23 | |
TYRGLVWXHJRKMT-UHFFFAOYNA-N | |
70832 | |
CC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
C11H13NO4 | |
MFCD00063126 | |
n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine | |
2-{[(benzyloxy)carbonyl]amino}propanoic acid |
Specifications
26607-51-2 | |
White | |
MFCD00063126 | |
n-cbz-d-alanine, z-d-ala-oh, cbz-d-ala-oh, z-d-alanine, n-benzyloxycarbonyl-d-alanine, n-carbobenzoxy-d-alanine, r-2-benzyloxy carbonyl amino propanoic acid, n-carbobenzyloxy-d-alanine, benzyloxycarbonyl-d-alanine, cbz-d-alanine | |
CC(NC(=O)OCC1=CC=CC=C1)C(O)=O | |
223.23 | |
223.23 | |
Crystalline Powder |
85°C | |
C11H13NO4 | |
1 g | |
TYRGLVWXHJRKMT-UHFFFAOYNA-N | |
2-{[(benzyloxy)carbonyl]amino}propanoic acid | |
70832 | |
≥98.0% (HPLC,T) | |
N-Carbobenzoxy-D-alanine |
Safety and Handling
TSCA : No