missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-1,12-diaminododecane Hydrochloride, TCI America™

Manufacturer:  TCI America C34725G

 View more versions of this product

Catalog No. C34725G



Specifications

Specifications

N-Carbobenzoxy-1,12-diaminododecane Hydrochloride
175°C
1051420-16-6
MFCD08272292
YISCQBUBEASQMD-UHFFFAOYSA-N
benzyl N-(12-aminododecyl)carbamate;hydrochloride
131865264
Crystal-Powder at 20°C
White
5g
C20H35ClN2O2
N-Cbz-1,12-diaminododecane Hydrochloride, Benzyl N-(12-Aminododecyl)carbamate Hydrochloride, N-(12-Aminododecyl)carbamic Acid Benzyl Ester Hydrochloride
C1=CC=C(C=C1)COC(=O)NCCCCCCCCCCCCN.Cl
370.962
370.96
≥98.0% (HPLC)

Chemical Identifiers

1051420-16-6
370.962
YISCQBUBEASQMD-UHFFFAOYSA-N
131865264
C1=CC=C(C=C1)COC(=O)NCCCCCCCCCCCCN.Cl
C20H35ClN2O2
MFCD08272292
N-Cbz-1,12-diaminododecane Hydrochloride, Benzyl N-(12-Aminododecyl)carbamate Hydrochloride, N-(12-Aminododecyl)carbamic Acid Benzyl Ester Hydrochloride
benzyl N-(12-aminododecyl)carbamate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents