missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbobenzoxy-1,10-diaminodecane Hydrochloride, TCI America™

Manufacturer:  TCI America C34711G

 View more versions of this product

Catalog No. C34711G



Specifications

Specifications

N-Carbobenzoxy-1,10-diaminodecane Hydrochloride
176°C
1051420-13-3
MFCD08272290
XDIZWAPVTINSPI-UHFFFAOYSA-N
benzyl N-(10-aminodecyl)carbamate;hydrochloride
131865263
Crystal-Powder at 20°C
White
1g
C18H31ClN2O2
N-Cbz-1,10-diaminodecane Hydrochloride, Benzyl N-(10-Aminodecyl)carbamate Hydrochloride, N-(10-Aminodecyl)carbamic Acid Benzyl Ester Hydrochloride
C1=CC=C(C=C1)COC(=O)NCCCCCCCCCCN.Cl
342.908
342.91
≥98.0% (HPLC)

Chemical Identifiers

1051420-13-3
342.908
XDIZWAPVTINSPI-UHFFFAOYSA-N
131865263
C1=CC=C(C=C1)COC(=O)NCCCCCCCCCCN.Cl
C18H31ClN2O2
MFCD08272290
N-Cbz-1,10-diaminodecane Hydrochloride, Benzyl N-(10-Aminodecyl)carbamate Hydrochloride, N-(10-Aminodecyl)carbamic Acid Benzyl Ester Hydrochloride
benzyl N-(10-aminodecyl)carbamate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents