Please login to your online account to display your discounted pricing

N-Carbethoxyphthalimide, 99+%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC108310100

 View more versions of this product

Catalog No. AC108310100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥99.0% (saponification)
Formula Weight 219.2
Quantity 10g
Color White
Beilstein 21, II, 357
Melting Point 82° to 84°C
Fieser 01,111; 05,91; 09,212
CAS Max % 100.0
CAS Min % 99.0
Packaging Glass bottle
Chemical Name or Material N-Carbethoxyphthalimide
Assay Percent Range 99+%
MDL Number MFCD00005893
Synonym ethyle1, 3-dioxo-2-isoindolinecarboxylate
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Molecular Formula C11H9NO4
CAS 22509-74-6
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-Carbethoxyphthalimide
CAS 22509-74-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0% (saponification)
Assay Percent Range 99+%
Molecular Formula C11H9NO4
Formula Weight 219.2
MDL Number MFCD00005893
Synonym ethyle1, 3-dioxo-2-isoindolinecarboxylate
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents