missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbethoxy-4-tropinone 98%, ACROS Organics™

$431.23 - $431.23

Chemical Identifiers

CAS 32499-64-2
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00078171
InChI Key ANEJUHJDPGTVIO-UHFFFAOYSA-N
Synonym 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, n-carbethoxynortropinone, n-ethoxycarbonyl nortropinone
PubChem CID 401882
IUPAC Name ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CCOC(=O)N1C2CCC1CC(=O)C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC134200250
SDS View Documents
Thermo Fisher Scientific
134200250
25g Glass Bottle Each for $431.23
 
Specifications

Chemical Identifiers

32499-64-2
197.234
ANEJUHJDPGTVIO-UHFFFAOYSA-N
401882
CCOC(=O)N1C2CCC1CC(=O)C2
C10H15NO3
MFCD00078171
3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, n-carbethoxynortropinone, n-ethoxycarbonyl nortropinone
ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate

Specifications

Authentic
1.4876 to 1.4896
25g
97.5
98%
MFCD00078171
ANEJUHJDPGTVIO-UHFFFAOYSA-N
ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
401882
Liquid
N-Carbethoxy-4-tropinone, 98%
Glass Bottle
Brown to Yellow
32499-64-2
100.0
C10H15NO3
3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, n-carbethoxynortropinone, n-ethoxycarbonyl nortropinone
CCOC(=O)N1C2CCC1CC(=O)C2
197.234
197.23
98%