Please login to your online account to display your discounted pricing

N-Carbethoxy-4-tropinone 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  134200250

 View more versions of this product

Catalog No. AC134200250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 197.23
Quantity 25g
Color Clear slightly yellow to light brown
CAS Max % 100.0
Synonym 3-Oxo-8-azabicylo[3.2.1]octane-8-carboxylic acid, ethyl ester
Infrared Spectrum Authentic
Refractive Index 1.4876 to 1.4896 (20°C, 589nm)
MDL Number MFCD00078171
Chemical Name or Material N-Carbethoxy-4-tropinone
Name Note 98%
Assay Percent Range 98%
CAS Min % 97.5
CAS 32499-64-2
Molecular Formula C10H15NO3
Packaging Glass bottle

Specifications

Chemical Name or Material N-Carbethoxy-4-tropinone
Name Note 98%
CAS 32499-64-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO3
Formula Weight 197.23
MDL Number MFCD00078171
Synonym 3-Oxo-8-azabicylo[3.2.1]octane-8-carboxylic acid, ethyl ester