missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbethoxy-4-piperidone, +98.5%, ACROS Organics™

Manufacturer:  Acros Organics 167240500

 View more versions of this product

Catalog No. AC167240500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Carbethoxy-4-piperidone
87°C
Glass bottle
0.5% max. (K.F.)
Colorless to Yellow
29976-53-2
100.0
C8H13NO3
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
CCOC(=O)N1CCC(=O)CC1
171.196
171.2
≥98.5%
1.1350g/mL
Authentic
1.4740 to 1.4760
95.0°C to 98.0°C (1.0mmHg)
50g
98.5
98.5+%
MFCD00006188
LUBGFMZTGFXIIN-UHFFFAOYSA-N
ethyl 4-oxopiperidine-1-carboxylate
34767
Liquid

Chemical Identifiers

29976-53-2
171.196
LUBGFMZTGFXIIN-UHFFFAOYSA-N
34767
CCOC(=O)N1CCC(=O)CC1
C8H13NO3
MFCD00006188
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
ethyl 4-oxopiperidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

Recommended Storage : Product may darken during storage

SDS Documents