missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Carbamoyl-DL-aspartic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1716618
Description
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.
Solubility
Soluble in hot water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications
∼175°C (decomposition) | |
923-37-5 | |
MFCD00042822 | |
ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid | |
HLKXYZVTANABHZ-UHFFFAOYSA-N | |
2-(carbamoylamino)butanedioic acid | |
279 | |
176.13 | |
50 g |
N-Carbamoyl-DL-aspartic acid | |
C5H8N2O5 | |
1726861 | |
Soluble in hot water. | |
C(C(C(=O)O)NC(=O)N)C(=O)O | |
176.128 | |
CHEBI:64850 | |
98% |
Chemical Identifiers
923-37-5 | |
176.128 | |
HLKXYZVTANABHZ-UHFFFAOYSA-N | |
279 | |
2-(carbamoylamino)butanedioic acid |
C5H8N2O5 | |
MFCD00042822 | |
ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid | |
CHEBI:64850 | |
C(C(C(=O)O)NC(=O)N)C(=O)O |
Safety and Handling
EINECSNumber : 213-096-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only