missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyrophenone p-toluenesulfonylhydrazone, 97%

Manufacturer:  Alfa Aesar™ H5342806

 View more versions of this product

Catalog No. AAH5342806



Specifications

Specifications

n-Butyrophenone p-toluenesulfonylhydrazone
5g
C17H20N2O2S
4-methyl-n-1-phenylhexan-2-ylideneamino benzenesulfonamide, 4-methyl-n'-2e-1-phenylbutan-2-ylidene benzenesulfonohydrazide
CCC(=NNS(=O)(=O)C1=CC=C(C=C1)C)CC2=CC=CC=C2
316.419
316.42
121°C to 125°C
41780-81-8
MFCD01230699
DQAAEKVIAXFJEK-FBMGVBCBSA-N
4-methyl-N-[(E)-1-phenylbutan-2-ylideneamino]benzenesulfonamide
77520436
97%

Chemical Identifiers

41780-81-8
316.419
DQAAEKVIAXFJEK-FBMGVBCBSA-N
77520436
CCC(=NNS(=O)(=O)C1=CC=C(C=C1)C)CC2=CC=CC=C2
C17H20N2O2S
MFCD01230699
4-methyl-n-1-phenylhexan-2-ylideneamino benzenesulfonamide, 4-methyl-n'-2e-1-phenylbutan-2-ylidene benzenesulfonohydrazide
4-methyl-N-[(E)-1-phenylbutan-2-ylideneamino]benzenesulfonamide
SDS Documents