Learn More
n-Butyronitrile, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 108140025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
n-Butyronitrile | |
98.5 | |
0.7900g/mL | |
16°C | |
98.5% min. (GC) | |
C4H7N | |
CH3CH2CH2CN | |
2.5L | |
0.79 | |
butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile | |
KVNRLNFWIYMESJ-UHFFFAOYSA-N | |
butanenitrile | |
8008 | |
CHEBI:51937 | |
99% |
109-74-0 | |
100.0 | |
115.0°C to 117.0°C | |
Authentic | |
Glass bottle | |
1.3830 to 1.3850 | |
MFCD00001968 | |
02, II, 252 | |
15, 1601 | |
Solubility in water: 33g/L (25°C). Other solubilities: miscible with alcohol,ether and dimethylformamide | |
CCCC#N | |
69.11 | |
0.57 mPa.s (20°C) | |
69.11 |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-700-6
RUO â Research Use Only