missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butyltin trichloride, 96%

Manufacturer:  Alfa Aesar™ 7112514

 View more versions of this product

Catalog No. AA7112514



Specifications

Specifications

n-Butyltin trichloride
-63°C
25g
Acrid
1.5244
C4H9Cl3Sn
UN3265
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyl(trichloro)stannane
14236
Liquid
98°C (10mmHg)
1118-46-3
81°C (178°F)
1.693g/mL
Moisture Sensitive
MFCD00000515
butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
CCCC[Sn](Cl)(Cl)Cl
282.176
282.17
96%

Chemical Identifiers

1118-46-3
282.176
YMLFYGFCXGNERH-UHFFFAOYSA-K
14236
CCCC[Sn](Cl)(Cl)Cl
C4H9Cl3Sn
MFCD00000515
butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
butyl(trichloro)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H372-H314-H318-H312-H227
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : WH6780000

TSCA : TSCA

SDS Documents