Please login to your online account to display your discounted pricing

Alfa Aesar™ n-(Butylthio)acetonitrile, 95%

Chemical Identifiers

Chemical Name or Material n-(Butylthio)acetonitrile
CAS 71037-08-6
Molecular Formula C6H11NS
Linear Formula CH3(CH2)3SCH2CN
Formula Weight 129.23
MDL Number MFCD00041458
UN Number UN3276
Beilstein 4174357
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2233606

  SDS
alfa aesar™
B2233606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2233614

  SDS
alfa aesar™
B2233614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.23
Health Hazard 1 Danger
Beilstein 4174357
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00041458
Assay 95%
Quantity 5g
UN Number UN3276
Molecular Formula C6H11NS
Boiling Point 134° to 135°C (40mmHg)
Linear Formula CH3(CH2)3SCH2CN
CAS 71037-08-6
Chemical Name or Material n-(Butylthio)acetonitrile
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.