missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butylmagnesium chloride, 20 wt.% solution in THF/Toluene, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 331681000

 View more versions of this product

Catalog No. AC331681000


 

Specifications Description & Specifications

Specifications

Solution
100mL
AcroSeal™ Glass Bottle in Tin Can
78.0,78.0,19.0
108-88-3,693-04-9,109-99-9
C4H9ClMg
Product may develop some turbidity or precipitate
116.87
Green-brown to brown
19 to 22% (as BUMC)
22.0,81.0,81.0
CH3(CH2)3MgCl
n-Butylmagnesium chloride

Chemical Identifiers

693-04-9,109-99-9,108-88-3
22.0,81.0,81.0
CH3(CH2)3MgCl
n-Butylmagnesium chloride
19.0,78.0,78.0
C4H9ClMg
116.87
19 to 22% (as BUMC)

Safety and Handling

SDS & Safety Documents

GHS P Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Suspected of damaging the unborn child. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container. Store in a dry place. Store in a closed container.

Danger