Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Butylmagnesium chloride, 1.5-2.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Grignard reagent

Chemical Identifiers

Chemical Name or Material n-Butylmagnesium chloride
CAS 693-04-9
Linear Formula CH3(CH2)3MgCl
Formula Weight 116.87
MDL Number MFCD00000475
UN Number UN3399
Beilstein 3587228
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41676AK

 
alfa aesar™
41676AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41676AD

 
alfa aesar™
41676AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.87
Chemical Name or Material n-Butylmagnesium chloride
TSCA TSCA
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive
Physical Form Liquid
Linear Formula CH3(CH2)3MgCl
Concentration 1.5 to 2.5M in THF
MDL Number MFCD00000475
CAS 693-04-9
UN Number UN3399
Beilstein 3587228
Color Brown
Quantity 250mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

  • Packaged under Argon