Please login to your online account to display your discounted pricing

n-Butyllithium, 2.6M solution in toluene, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC378938000

 View more versions of this product

Catalog No. AC378938000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 64.06
Quantity 800mL
Color Yellow to orange
Packaging AcroSeal™ Glass Bottle
Assay 19.8 to 20.2% (as BuLi)
Density 0.8600g/mL
Linear Formula CH3(CH2)3Li
Free Alkali 0.4% max. (as BuLi)
Additional Information product may develop some turbidity or precipitate
Chemical Name or Material n-Butyllithium
MDL Number MFCD00009414
Health Hazard 1 Danger
CAS 108-88-3
Name Note 2.6M solution in toluene
Molecular Formula C4H9Li
CAS 109-72-8
Health Hazard 2 GHS P Statement May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Specifications

Chemical Name or Material n-Butyllithium
Name Note 2.6M solution in toluene
CAS 109-72-8
CAS 108-88-3
Assay 19.8 to 20.2% (as BuLi)
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
Formula Weight 64.06
MDL Number MFCD00009414
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of

SDS & Safety Documents