Learn More
n-Butyllithium, 2.7M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 378931000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Butyllithium | |
2.6M solution in toluene | |
C4H9Li | |
MFCD00009414 | |
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc | |
MZRVEZGGRBJDDB-UHFFFAOYSA-N | |
butyllithium | |
61028 | |
19.8 to 20.2% (as BuLi) | |
0.86 | |
4°C |
Yellow to Orange | |
109-72-8,108-88-3 | |
CH3(CH2)3Li | |
0.8600g/mL | |
Solubility in water: vigorous reaction. | |
[Li]CCCC | |
64.06 | |
64.06 | |
AcroSeal™ Glass bottle | |
100 mL | |
Solution |
Chemical Identifiers
109-72-8 | |
64.06 | |
MZRVEZGGRBJDDB-UHFFFAOYSA-N | |
61028 | |
[Li]CCCC |
C4H9Li | |
MFCD00009414 | |
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc | |
butyllithium |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Danger
RUO – Research Use Only