Please login to your online account to display your discounted pricing

n-Butyllithium, 2.2M solution in Cyclohexane, AcroSeal™, ACROS Organics™

$54.62 - $259.75

Chemical Identifiers

Chemical Name or Material n-Butyllithium
Name Note 2.2M Solution in Cyclohexane
CAS 109-72-8
CAS 110-82-7
CAS 110-54-3
Assay 19.5 to 20.5% (as BuLi) (Active base)
Molecular Formula C4H9Li
Linear Formula CH3(CH2)3Li
Formula Weight 64.06
MDL Number MFCD00009414
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377491000

  SDS
thermo fisher scientific
AC377491000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $54.62

AC377498000

  SDS
thermo fisher scientific
AC377498000
AcroSeal™ Glass Bottle 800mL Each for $259.75
Description & Specifications

Specifications

Assay 19.5 to 20.5% (as BuLi) (Active base)
Color Grey to brown
Physical Form Solution, magnesium salts may crystallize below 20°C
Formula Weight 64.06
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.7500g/mL
Linear Formula CH3(CH2)3Li
MDL Number MFCD00009414
Health Hazard 1 GHS Signal Word: Danger
Free Alkali 1 % max. (as BuLi)
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Suspected of damaging fertility.
Catches fire spontaneously if exposed to air.
CAS 110-82-7
CAS 110-54-3
Chemical Name or Material n-Butyllithium
Name Note 2.2M Solution in Cyclohexane
Molecular Formula C4H9Li
CAS 109-72-8
Additional Information product may develop some turbidity or precipitate
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Suspected of damaging fertility.
Catches fire spontaneously if exposed to air.